Please login to your online account to display your discounted pricing

Alfa Aesar™ Diphenyliodonium nitrate, 97%

Chemical Identifiers

Chemical Name or Material Diphenyliodonium nitrate
CAS 722-56-5
Molecular Formula C12H10INO3
Formula Weight 343.12
MDL Number MFCD00031704
UN Number UN3087
Beilstein 3922747
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2186103

 
alfa aesar™
B2186103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2186106

 
alfa aesar™
B2186106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 343.12
Chemical Name or Material Diphenyliodonium nitrate
Odor Odorless
Assay 97%
Molecular Formula C12H10INO3
MDL Number MFCD00031704
CAS 722-56-5
UN Number UN3087
Beilstein 3922747
Melting Point ≈150°C (dec.)
Health Hazard 1 Danger
Quantity 1g
Health Hazard 2 GHS H Statement
H272-H301-H315-H319-H335
May intensify fire; oxidizer.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P221-P210-P301+P310-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S.
RTECS Number NN6665000
Safety and Handling

Danger

GHS P Statement
P221-P210-P301+P310-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H272-H301-H315-H319-H335
May intensify fire; oxidizer.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.