Please login to your online account to display your discounted pricing

Alfa Aesar™ Diphenyliodonium iodide, 98%

$108.34 - $1,448.00

Chemical Identifiers

Chemical Name or Material Diphenyliodonium iodide
CAS 2217-79-0
Molecular Formula C12H10I2
Formula Weight 408.02
MDL Number MFCD00011910
Beilstein 3918215
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1943806

  SDS
alfa aesar™
A1943806
5g Each for $108.34

AAA1943814

  SDS
alfa aesar™
A1943814
25g Each for $417.38

AAA1943822

  SDS
alfa aesar™
A1943822
100g Each for $1,448.00
Description & Specifications

Specifications

Formula Weight 408.02
Chemical Name or Material Diphenyliodonium iodide
Assay 98%
Molecular Formula C12H10I2
MDL Number MFCD00011910
CAS 2217-79-0
Beilstein 3918215
Melting Point 162° to 164°C
Health Hazard 1 Warning
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.