Please login to your online account to display your discounted pricing

Alfa Aesar™ Diphenyliodonium hexafluorophosphate, 98%

Chemical Identifiers

Chemical Name or Material Diphenyliodonium hexafluorophosphate
CAS 58109-40-3
Formula Weight 426.08g/mol
MDL Number MFCD00061398
UN Number UN2811
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1890606

 
alfa aesar™
18906-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1890614

 
alfa aesar™
18906-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 5g
Melting Point 138° to 140°C
Quantity 5g
Chemical Name or Material Diphenyliodonium hexafluorophosphate
CAS 58109-40-3
Physical Form Crystalline
Assay 98%
Formula Weight 426.08g/mol
MDL Number MFCD00061398
UN Number UN2811
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA TSCA