Please login to your online account to display your discounted pricing

Diphenyliodonium Bromide 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC408950050

 View more versions of this product

Catalog No. AC408950050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 361.01
Physical Form Powder
Quantity 5g
Color Light yellow
TSCA TSCA
EINECS Number 216-050-3
Melting Point 224° to 228°C
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Toxic if swallowed.
Name Note 98%
Molecular Formula C12H10I·Br
MDL Number MFCD00031703
Linear Formula (C6H5)2I+Br-
CAS 1483-73-4
Assay Percent Range 98%
Chemical Name or Material Diphenyliodonium bromide

Specifications

Chemical Name or Material Diphenyliodonium bromide
Name Note 98%
CAS 1483-73-4
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12H10I·Br
Linear Formula (C6H5)2I+Br-
Formula Weight 361.01
MDL Number MFCD00031703
Accessories

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Toxic if swallowed.

SDS & Safety Documents