Please login to your online account to display your discounted pricing

Diphenyliodonium Bromide 98%, ACROS Organics™

$308.01 - $308.01

Chemical Identifiers

Chemical Name or Material Diphenyliodonium bromide
Name Note 98%
CAS 1483-73-4
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12H10I·Br
Linear Formula (C6H5)2I+Br-
Formula Weight 361.01
MDL Number MFCD00031703
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC408950250

  SDS
thermo fisher scientific
408950250
Glass bottle 25g Each for $308.01

AC408950050

  SDS
thermo fisher scientific
AC408950050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder
Formula Weight 361.01
Quantity 25g
Color Light yellow
EINECS Number 216-050-3
TSCA TSCA
Melting Point 224° to 228°C
Chemical Name or Material Diphenyliodonium bromide
Infrared Spectrum Authentic
Name Note 98%
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C12H10I·Br
Linear Formula (C6H5)2I+Br-
MDL Number MFCD00031703
CAS 1483-73-4
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Toxic if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Toxic if swallowed.