Please login to your online account to display your discounted pricing

Alfa Aesar™ Diphenyliodonium bromide, 98%

Chemical Identifiers

Chemical Name or Material Diphenyliodonium bromide
CAS 1483-73-4
Molecular Formula C12H10BrI
Formula Weight 361.03g/mol
MDL Number MFCD00031703
UN Number UN2811
Beilstein 3918437
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1299609

 
alfa aesar™
A12996-09
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Odor Odorless
Melting Point ≈244°C (subl.)
Quantity 10g
Chemical Name or Material Diphenyliodonium bromide
CAS 1483-73-4
Assay 98%
Molecular Formula C12H10BrI
Formula Weight 361.03g/mol
MDL Number MFCD00031703
UN Number UN2811
Beilstein 3918437
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.