Please login to your online account to display your discounted pricing

Diphenylglyoxime, 97%


Manufacturer: thermo fisher scientific  AC408931000

 View more versions of this product

Catalog No. AC408931000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Formula Weight 240.26
Quantity 100g
Chemical Name or Material Diphenylglyoxime
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C14H12N2O2
MDL Number MFCD00002113
CAS 23873-81-6

Specifications

Chemical Name or Material Diphenylglyoxime
CAS 23873-81-6
Purity Grade Technical Grade
Molecular Formula C14H12N2O2
Formula Weight 240.26
MDL Number MFCD00002113