Please login to your online account to display your discounted pricing

Diphenylglyoxime, 97%

Chemical Identifiers

Chemical Name or Material Diphenylglyoxime
CAS 23873-81-6
Purity Grade Technical Grade
Molecular Formula C14H12N2O2
Formula Weight 240.26
MDL Number MFCD00002113
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

AC408930250

  SDS
thermo fisher scientific
AC408930250
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC408931000

  SDS
thermo fisher scientific
AC408931000
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 240.26
Purity Grade Technical Grade
Quantity 25g
CAS 23873-81-6
Assay Percent Range 97%
Chemical Name or Material Diphenylglyoxime
MDL Number MFCD00002113
Molecular Formula C14H12N2O2