Please login to your online account to display your discounted pricing

Diphenyleneiodonium chloride, Tocris


Binds to flavoproteins and inhibits NOS

Quantity 50mg

Manufacturer: r & d systems  RDS050450

 View more versions of this product

Catalog No. 050450

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Diphenyleneiodonium chloride
CAS 4673-26-1
Recommended Storage Desiccate at -20°C
Assay Percent Range >99%
Molecular Formula C12H8ClI
Formula Weight 314.55
Synonym [1, 1'-Biphenyl]-2, 2'-diyliodonium chloride
Quantity 50mg

Specifications

Chemical Name or Material Diphenyleneiodonium chloride
Assay Percent Range >99%
Molecular Formula C12H8ClI
Formula Weight 314.55
Synonym [1, 1'-Biphenyl]-2, 2'-diyliodonium chloride
Accessories

Desiccate at -20°C