missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diphenyleneiodonium chloride, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J64838MF
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diphenyleneiodonium chloride | |
>310°C | |
C12H8ClI | |
50 mg | |
FCFZKAVCDNTYID-UHFFFAOYSA-M | |
314.55 | |
CHEBI:77967 | |
Powder |
4673-26-1 | |
White | |
MFCD00214165 | |
diphenyleneiodonium chloride, dibenziodolium chloride, unii-7m9d81yz2n, dpi, 8? 3-iodatricyclo 7.4.0.0 2 ,? trideca-1 13 ,2,4,6,9,11-hexaen-8-ylium chloride, dibenziodolium, chloride, d0n0gj, c12h8i.cl, dibenzo b,d iodolium chloride, 2,2'-biphenylyleneiodonium chloride | |
C1=CC=C2C(=C1)C3=CC=CC=C3[I+]2.[Cl-] | |
2733504 | |
314.55 |
Chemical Identifiers
4673-26-1 | |
314.55 | |
FCFZKAVCDNTYID-UHFFFAOYSA-M | |
2733504 | |
C1=CC=C2C(=C1)C3=CC=CC=C3[I+]2.[Cl-] |
C12H8ClI | |
MFCD00214165 | |
diphenyleneiodonium chloride, dibenziodolium chloride, unii-7m9d81yz2n, dpi, 8? 3-iodatricyclo 7.4.0.0 2 ,? trideca-1 13 ,2,4,6,9,11-hexaen-8-ylium chloride, dibenziodolium, chloride, d0n0gj, c12h8i.cl, dibenzo b,d iodolium chloride, 2,2'-biphenylyleneiodonium chloride | |
CHEBI:77967 |
Safety and Handling
P264b-P280i-P305+P351+P338
H319
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only