missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Diphenyldithiophosphonic acid

Manufacturer:  Alfa Aesar™ 1775809

 View more versions of this product

Catalog No. AA1775809



Specifications

Specifications

Diphenyldithiophosphonic acid
10g
C12H11PS2
diphenylphosphinodithioic acid, diphenyldithiophosphonic acid, diphenyldithiophosphinic acid, phosphinodithioic acid, diphenyl, diphenyl sulfanyl-??-phosphanethione, dithiodiphenylphosphinic acid, diphenyl-sulfanyl-sulfanylidene, diphenyl sulfanyl-lambda-phosphanethione, diphenyl-sulfanyl-sulfanylidenephosphorane, diphenyl sulfanyl-$l; 5-phosphanethione
C1=CC=C(C=C1)P(=S)(C2=CC=CC=C2)S
250.314
250.31
55°C to 56°C
1015-38-9
MFCD00014457
CLUOCCWZZAGLPM-UHFFFAOYSA-N
diphenyl-sulfanyl-sulfanylidene-$l^{5}-phosphane
129564
Crystalline

Chemical Identifiers

1015-38-9
250.314
CLUOCCWZZAGLPM-UHFFFAOYSA-N
129564
C1=CC=C(C=C1)P(=S)(C2=CC=CC=C2)S
C12H11PS2
MFCD00014457
diphenylphosphinodithioic acid, diphenyldithiophosphonic acid, diphenyldithiophosphinic acid, phosphinodithioic acid, diphenyl, diphenyl sulfanyl-??-phosphanethione, dithiodiphenylphosphinic acid, diphenyl-sulfanyl-sulfanylidene, diphenyl sulfanyl-lambda-phosphanethione, diphenyl-sulfanyl-sulfanylidenephosphorane, diphenyl sulfanyl-$l; 5-phosphanethione
diphenyl-sulfanyl-sulfanylidene-$l^{5}-phosphane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

SDS Documents