missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diphenyldichlorosilane, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1205136

 View more versions of this product

Catalog No. AAA1205136



Description

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuffs.

Solubility
Decomposes in water.

Notes
Moisture Sensitive. Store away from water/moisture, oxidizing agents and bases. Store under inert gas.
Specifications

Specifications

303°C to 304°C
-22°C
500 g
MFCD00000489
609882
diphenyldichlorosilane, silane, dichlorodiphenyl, dichloro diphenyl silane, diphenylsilicon dichloride, diphenylsilyl dichloride, dichlor-difenylsilan, diphenyl dichlorosilane, dichlor-difenylsilan czech, hsdb 316, benzene, 1,1'-dichlorosilylene bis
OSXYHAQZDCICNX-UHFFFAOYSA-N
dichloro(diphenyl)silane
6627
97%
1.579
1.208
Diphenyldichlorosilane
80-10-4
C12H10Cl2Si
UN1769
157°C (314°F)
Decomposes in water.
Cl[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1
253.20
253.2
Pungent
Moisture sensitive

Chemical Identifiers

80-10-4
253.20
OSXYHAQZDCICNX-UHFFFAOYSA-N
6627
Cl[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1
C12H10Cl2Si
MFCD00000489
diphenyldichlorosilane, silane, dichlorodiphenyl, dichloro diphenyl silane, diphenylsilicon dichloride, diphenylsilyl dichloride, dichlor-difenylsilan, diphenyl dichlorosilane, dichlor-difenylsilan czech, hsdb 316, benzene, 1,1'-dichlorosilylene bis
dichloro(diphenyl)silane
Safety and Handling

Safety and Handling

GHS H Statement
H310-H314-H318-H302
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P260-P262-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H310-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: DIPHENYLDICHLOROSILANE

EINECSNumber : 201-251-0

RTECSNumber : VV3190000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only