Learn More
Diphenyldichlorosilane, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1205122
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuffs.
Solubility
Decomposes in water.
Notes
Moisture Sensitive. Store away from water/moisture, oxidizing agents and bases. Store under inert gas.
Specifications
303°C to 304°C | |
-22°C | |
100 g | |
MFCD00000489 | |
609882 | |
diphenyldichlorosilane, silane, dichlorodiphenyl, dichloro diphenyl silane, diphenylsilicon dichloride, diphenylsilyl dichloride, dichlor-difenylsilan, diphenyl dichlorosilane, dichlor-difenylsilan czech, hsdb 316, benzene, 1,1'-dichlorosilylene bis | |
OSXYHAQZDCICNX-UHFFFAOYSA-N | |
dichloro(diphenyl)silane | |
6627 | |
97% | |
1.579 | |
1.208 |
Diphenyldichlorosilane | |
80-10-4 | |
C12H10Cl2Si | |
UN1769 | |
157°C (314°F) | |
Decomposes in water. | |
Cl[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1 | |
253.20 | |
253.2 | |
Pungent | |
Moisture sensitive |
Chemical Identifiers
80-10-4 | |
253.20 | |
OSXYHAQZDCICNX-UHFFFAOYSA-N | |
6627 | |
Cl[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1 |
C12H10Cl2Si | |
MFCD00000489 | |
diphenyldichlorosilane, silane, dichlorodiphenyl, dichloro diphenyl silane, diphenylsilicon dichloride, diphenylsilyl dichloride, dichlor-difenylsilan, diphenyl dichlorosilane, dichlor-difenylsilan czech, hsdb 316, benzene, 1,1'-dichlorosilylene bis | |
dichloro(diphenyl)silane |
Safety and Handling
GHS H Statement
H310-H314-H318-H302
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P262-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H310-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: DIPHENYLDICHLOROSILANE
EINECSNumber : 201-251-0
RTECSNumber : VV3190000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only