Learn More
Diphenylcyclopropenone, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 175820050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diphenylcyclopropenone | |
117.0°C to 121.0°C | |
Authentic | |
Glass bottle | |
MFCD00001311 | |
07,IV,1736 | |
diphenylcyclopropenone, diphencyprone, 2,3-diphenylcyclopropenone, 2,3-diphenylcycloprop-2-enone, cyclopropenone, diphenyl, dpcp, 2-cyclopropen-1-one, 2,3-diphenyl, 1,2-diphenylcyclopropen-3-one, cyclopropenone, 2,3-diphenyl, unii-i7g14nw5ec | |
HCIBTBXNLVOFER-UHFFFAOYSA-N | |
2,3-diphenylcycloprop-2-en-1-one | |
65057 | |
206.24 | |
Crystalline Powder |
886-38-4 | |
Beige to Orange-Pink | |
97.5% min. (HPLC) | |
C15H10O | |
5 g | |
15,3338 | |
Solubility in water: insoluble. Other solubilities: soluble in methanol, ethanol, acetone, chloroform,, dichloromethane, cyclohexane, benzene, hexane,, petroleum ether | |
O=C1C(=C1C1=CC=CC=C1)C1=CC=CC=C1 | |
206.24 | |
CHEBI:53074 | |
98% |
Chemical Identifiers
886-38-4 | |
206.24 | |
HCIBTBXNLVOFER-UHFFFAOYSA-N | |
65057 | |
2,3-diphenylcycloprop-2-en-1-one |
C15H10O | |
MFCD00001311 | |
diphenylcyclopropenone, diphencyprone, 2,3-diphenylcyclopropenone, 2,3-diphenylcycloprop-2-enone, cyclopropenone, diphenyl, dpcp, 2-cyclopropen-1-one, 2,3-diphenyl, 1,2-diphenylcyclopropen-3-one, cyclopropenone, 2,3-diphenyl, unii-i7g14nw5ec | |
CHEBI:53074 | |
O=C1C(=C1C1=CC=CC=C1)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
IF IN EYES: Rinse cautious
GHS Signal Word: Warning
EINECSNumber : 212-948-4
RUO – Research Use Only