Please login to your online account to display your discounted pricing

Diphenylcyclopropenone, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC175820250

 View more versions of this product

Catalog No. AC175820250

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Formula Weight 206.24
Color Slightly pinkish-orange to beige
Assay ≥97.5% (HPLC)
Quantity 25g
Melting Point 117° to 121°C
Chemical Name or Material Diphenylcyclopropenone
CAS 886-38-4
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 98%
Synonym DPC
Health Hazard 1 Warning
Molecular Formula C15H10O

Specifications

Chemical Name or Material Diphenylcyclopropenone
Assay ≥97.5% (HPLC)
Assay Percent Range 98%
Molecular Formula C15H10O
Formula Weight 206.24
Synonym DPC
Accessories

Warning

SDS & Safety Documents