Please login to your online account to display your discounted pricing

Diphenylcarbazone-Xylene Cyanol FF, For Chloride, Certified, LabChem™

$68.06 - $68.06

Chemical Identifiers

Chemical Name or Material Diphenylcarbazone-Xylene Cyanol FF
CAS 67-63-0
CAS Max % 99.72
CAS 538-62-5
CAS Max % 0.25
CAS 538-62-5
CAS 140-22-7
CAS 4463-44-9
CAS Max % 0.03
Name Note For Chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC136827

 
lab chem
LC136827
125mL Each for $68.06
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 242.28
Chemical Name or Material Diphenylcarbazone-Xylene Cyanol FF
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide; May release flammable gases; Sulfur compounds
CAS 140-22-7
Color Dark blue
Quantity 125mL
CAS 4463-44-9
CAS Max % 0.03
Name Note For Chloride
Physical Form Liquid
Linear Formula C13H14N4O
CAS 67-63-0
UN Number UN1993
Health Hazard 1 Danger
CAS Max % 99.72
Health Hazard 2 GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Identification Passes Test
CAS 538-62-5
Health Hazard 3 GHS P Statement
Keep away from heat, hot surfaces, open flames, sparks, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Avoid breathing vapors.
Use only outdoors or in a well-ventilated area.
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Packaging Amber Glass
CAS Max % 0.25
Solubility Soluble in water
CAS 538-62-5
Suitability Passes Test
Storage Information Refrigerate
Safety and Handling

Danger

GHS P Statement
Keep away from heat, hot surfaces, open flames, sparks, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Avoid breathing vapors.
Use only outdoors or in a well-ventilated area.
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.