Please login to your online account to display your discounted pricing

Diphenylcarbazone TS, 1% (w/v) Alcoholic Solution, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Diphenylcarbazone
Name Note Test Solution
CAS 64-17-5
CAS Min % 97.0
CAS Max % 100.0
CAS 538-62-5
CAS Min % 1.21
CAS Max % 1.26
CAS 67-56-1 (Methanol (Methyl Alcohol))
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

2605-4

 
ricca chemical company
RI26054
Glass amber 120mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear red
Quantity 120mL
Chemical Name or Material Diphenylcarbazone
CAS 64-17-5
Name Note Test Solution
Solvent or Matrix Alcoholic Solution
CAS Min % 1.21
Concentration or Composition Notes 1% (w/v)
CAS Min % 97.0
CAS Max % 100.0
CAS Max % 1.26
Packaging Glass amber
CAS 67-56-1 (Methanol (Methyl Alcohol))
CAS 538-62-5