Please login to your online account to display your discounted pricing

Diphenylcarbazone-Bromophenol Blue Mixed Indicator, for Chloride Determination, Ricca Chemical


Quantity 4L
Packaging Glass amber

Manufacturer: ricca chemical company  RIR26100004C

 View more versions of this product

Catalog No. R26100004C

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear dark red
Quantity 4L
Chemical Name or Material Diphenylcarbazone-Bromophenol Blue Mixed Indicator
CAS Max % 0.06
CAS 62625-28-9 (Bromophenol Blue, Sodium Salt)
Packaging Glass amber
CAS 64-17-5
CAS Min % 97.0
CAS Max % 100.0
CAS 538-62-5
CAS Min % 0.61
CAS Max % 0.63
CAS 62625-28-9
CAS Min % 0.06

Specifications

Chemical Name or Material Diphenylcarbazone-Bromophenol Blue Mixed Indicator
CAS 64-17-5
CAS Min % 97.0
CAS Max % 100.0
CAS 538-62-5
CAS Min % 0.61
CAS Max % 0.63
CAS 62625-28-9
CAS Min % 0.06
CAS Max % 0.06
CAS 62625-28-9 (Bromophenol Blue, Sodium Salt)
Physical Form Liquid