Please login to your online account to display your discounted pricing

Diphenylcarbazide, ACS Grade, LabChem™

$174.94 - $174.94

Chemical Identifiers

Chemical Name or Material Diphenylcarbazide
CAS 140-22-7
CAS Max % 100
Purity Grade ACS Grade
Linear Formula C13H14N4O
Formula Weight 242.28
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC136558

 
lab chem
LC136558
Amber Glass 25g Each for $174.94
Description & Specifications

Specifications

Formula Weight 242.28
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Chemical Name or Material Diphenylcarbazide
Identification Passes Test
CAS Max % 100
Storage Information Room Temperature
Color Off-white
Packaging Amber Glass
Quantity 25g
Health Hazard 3 GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Linear Formula C13H14N4O
Purity Grade ACS Grade
Physical Form Powder
CAS 140-22-7
Safety and Handling

Warning

GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.