Please login to your online account to display your discounted pricing

Diphenylcarbazide, 1% Alcoholic, Certified, LabChem™

$59.86 - $59.86

Chemical Identifiers

Chemical Name or Material Diphenylcarbazide
CAS 64-17-5
CAS Max % 89.1
CAS 67-63-0
CAS Max % 4.95
CAS 67-56-1
CAS Max % 4.95
CAS 140-22-7
CAS Max % 1
Name Note 1% Alcoholic
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC136607

 
lab chem
LC136607
White Poly 125mL Each for $59.86
Description & Specifications

Specifications

Formula Weight 242.28
Purity Grade Certified Grade
Decomposition Information Carbon monoxide; Carbon dioxide; May release flammable gases
Chemical Name or Material Diphenylcarbazide
CAS Max % 4.95
Identification Passes Test
CAS 140-22-7
CAS Max % 1
Name Note 1% Alcoholic
Packaging White Poly
Physical Form Liquid
Linear Formula C13H14N4O
Solubility Soluble in water
CAS 64-17-5
Storage Information Room Temperature
UN Number UN1170
Melting Point −115°C
Boiling Point 78°C
Health Hazard 1 Danger
Color Clear, colorless to slight yellow
Quantity 125mL
Flash Point 13°C
CAS Max % 89.1
Health Hazard 2 GHS H Statement
Highly flammable liquid and vapor.
Suspected of damaging fertility or the unborn child.
Causes damage to organs (central nervous system, optic nerve) (oral, dermal).
CAS 67-63-0
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, spray, vapors.
Do not eat, drink or smoke when using this product.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
In case of fire: Use carbon dioxide, powder, alcohol-resistant foam to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Specific Gravity 790kg/m3
CAS Max % 4.95
Density 790kg/m3
Suitability Passes Test
CAS 67-56-1
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, spray, vapors.
Do not eat, drink or smoke when using this product.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
In case of fire: Use carbon dioxide, powder, alcohol-resistant foam to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Highly flammable liquid and vapor.
Suspected of damaging fertility or the unborn child.
Causes damage to organs (central nervous system, optic nerve) (oral, dermal).