Learn More
Diphenylborinic acid 2-aminoethyl ester, 98%, Thermo Scientific Chemicals
$132.37 - $132.37
Chemical Identifiers
CAS | 524-95-8 |
---|---|
Molecular Formula | C14H16BNO |
Molecular Weight (g/mol) | 225.10 |
MDL Number | MFCD00014823 |
InChI Key | BLZVCIGGICSWIG-UHFFFAOYSA-N |
Synonym | 2-aminoethyl diphenylborinate, 2-apb, 2-aminoethoxy diphenylborane, 2-aminoethoxydiphenylborane, 2-aminoethoxydiphenylborate, 2-aminoethoxydiphenyl borate, o-2-aminoethyl diphenylborinic acid, b-2-aminoethoxy diphenylborane, 2-diphenylboryl oxy ethanamine, borinic acid, o-2-aminoethyl diphenyl |
PubChem CID | 1598 |
SMILES | NCCOB(C1=CC=CC=C1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC155420050
|
Thermo Scientific Chemicals
155420050 |
5 g | Glass bottle |
Each for $132.37
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
524-95-8 | |
225.10 | |
BLZVCIGGICSWIG-UHFFFAOYSA-N | |
1598 |
C14H16BNO | |
MFCD00014823 | |
2-aminoethyl diphenylborinate, 2-apb, 2-aminoethoxy diphenylborane, 2-aminoethoxydiphenylborane, 2-aminoethoxydiphenylborate, 2-aminoethoxydiphenyl borate, o-2-aminoethyl diphenylborinic acid, b-2-aminoethoxy diphenylborane, 2-diphenylboryl oxy ethanamine, borinic acid, o-2-aminoethyl diphenyl | |
NCCOB(C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
524-95-8 | |
Colorless to Yellow | |
98% | |
C14H16BNO | |
MFCD00014823 | |
2-aminoethyl diphenylborinate, 2-apb, 2-aminoethoxy diphenylborane, 2-aminoethoxydiphenylborane, 2-aminoethoxydiphenylborate, 2-aminoethoxydiphenyl borate, o-2-aminoethyl diphenylborinic acid, b-2-aminoethoxy diphenylborane, 2-diphenylboryl oxy ethanamine, borinic acid, o-2-aminoethyl diphenyl | |
BLZVCIGGICSWIG-UHFFFAOYSA-N | |
2-diphenylboranyloxyethanamine | |
1598 | |
98% | |
Diphenylborinic acid 2-aminoethyl ester, 98% |
190.0°C to 194.0°C | |
Authentic | |
Glass bottle | |
(C6H5)2BOCH2CH2NH2 | |
5 g | |
Solubility in water: insoluble. Other solubilities: soluble in ethanol,methanol | |
NCCOB(C1=CC=CC=C1)C1=CC=CC=C1 | |
225.10 | |
225.1 | |
Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and
GHS Signal Word: Warning
EINECSNumber : 208-366-5
RTECSNumber : ED6150000
RUO – Research Use Only