missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diphenylamine, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Manufacturer:  Avantor Performance 194401

 View more versions of this product

Catalog No. 02002752



Specifications

Specifications

Diphenylamine
Wide Mouth Amber Glass Bottle
500g
C12H11N
DMBHHRLKUKUOEG-UHFFFAOYSA-N
N-phenylaniline
11487
169.23
BAKER ANALYZED™ ACS Reagent
150.00°C
1L = 1.16kg
122-39-4
diphenylamine, n-phenylbenzenamine, anilinobenzene, benzenamine, n-phenyl, n,n-diphenylamine, phenylaniline, scaldip, n-phenylbenzeneamine, big dipper, no-scald
C1=CC=C(C=C1)NC2=CC=CC=C2
169.227
CHEBI:4640
Crystals

Chemical Identifiers

122-39-4
169.227
diphenylamine, n-phenylbenzenamine, anilinobenzene, benzenamine, n-phenyl, n,n-diphenylamine, phenylaniline, scaldip, n-phenylbenzeneamine, big dipper, no-scald
CHEBI:4640
C1=CC=C(C=C1)NC2=CC=CC=C2
C12H11N
DMBHHRLKUKUOEG-UHFFFAOYSA-N
11487
N-phenylaniline
Documents