Please login to your online account to display your discounted pricing

Diphenylamine (Certified ACS), Fisher Chemical

$272.31 - $896.77

Chemical Identifiers

Chemical Name or Material Diphenylamine
CAS 122-39-4
Purity Grade Certified ACS Grade
Physical Form Solid
Molecular Formula C12H11N
Linear Formula (C6H5)2NH
Formula Weight 169.22
MDL Number MFCD00003014
Synonym N-Phenylbenzeneamine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

O2611-100

  SDS
fisher scientific company
O2611100
Glass Bottle 100g Each for $272.31
Case of 6 Each for $1,633.86

O2611-500

  SDS
fisher scientific company
O2611500
Poly Bottle 500g Each for $896.77
Case of 6 Each for $5,380.62
Description & Specifications

Specifications

Melting Point 52°C
Boiling Point 302°C
Formula Weight 169.22
Purity Grade Certified ACS Grade
Quantity 100g
DOT Information DOT Class 6.1, : Poison
Physical Form Solid
Molecular Formula C12H11N
Color White Yellow Brown
Nitrate Pass Test
Identification Pass Test
Additional Information Vapor Pressure: 0.0003hPa at 20°C
MDL Number MFCD00003014
ChemAlert Storage Symbol Blue
Packaging Glass Bottle
Solubility in Alcohol Pass Test
Ignition Residue 0.03% max.
Sensitivity Pass Test
Synonym N-Phenylbenzeneamine
Chemical Name or Material Diphenylamine
CAS 122-39-4
Linear Formula (C6H5)2NH
Safety and Handling