Please login to your online account to display your discounted pricing

Diphenylamine, Approx. 99%, MP Biomedicals™


Quantity 5g

Manufacturer: mp biomedicals  0215012405

 View more versions of this product

Catalog No. ICN15012405

  • / Each
 

Description & Specifications

Chemical Identifiers

Appearance Dark Taupe Flakes
Density 1.16g/cm3 at 20°C (L)
Odor Pleasant floral odor
Quantity 5g
Chemical Name or Material Diphenylamine
CAS 122-39-4
Physical Form Solid
Assay Percent Range Approx. 99%
Molecular Formula C12H11N
Formula Weight 169.23g/mol
MDL Number MFCD00003014
Synonym N-phenylaniline
Storage Note 1 Protect from light.
Recommended Storage Store at Room Temperature (15deg to 30°C)
EINECS Number 204-539-4

Specifications

Chemical Name or Material Diphenylamine
Assay Percent Range Approx. 99%
Molecular Formula C12H11N
Formula Weight 169.23g/mol
MDL Number MFCD00003014
Synonym N-phenylaniline
Accessories

Store at Room Temperature (15deg to 30°C)

Protect from light.