Please login to your online account to display your discounted pricing

Diphenylamine, Approx. 99%, MP Biomedicals™

$45.66 - $45.66

Chemical Identifiers

Chemical Name or Material Diphenylamine
Assay Percent Range Approx. 99%
Molecular Formula C12H11N
Formula Weight 169.23g/mol
MDL Number MFCD00003014
Synonym N-phenylaniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN15012405

 
mp biomedicals
0215012405
5g Each for $45.66
Description & Specifications

Specifications

Appearance Dark Taupe Flakes
Density 1.16g/cm3 at 20°C (L)
Odor Pleasant floral odor
Quantity 5g
Chemical Name or Material Diphenylamine
CAS 122-39-4
Physical Form Solid
Assay Percent Range Approx. 99%
Molecular Formula C12H11N
Formula Weight 169.23g/mol
MDL Number MFCD00003014
Synonym N-phenylaniline
Storage Note 1 Protect from light.
Recommended Storage Store at Room Temperature (15deg to 30°C)
EINECS Number 204-539-4
Safety and Handling

Store at Room Temperature (15deg to 30°C)

Protect from light.