Please login to your online account to display your discounted pricing

Diphenylamine ACS, Ricca Chemical


Quantity 100g
Packaging Poly wht WM

Manufacturer: ricca chemical company  RIRDCD0650100B1

 View more versions of this product

Catalog No. RDCD0650100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 100g
Purity Grade ACS Grade
Chemical Name or Material Diphenylamine
CAS Max % 100.0
CAS Min % 98.0
Shelf Life 60 months
Packaging Poly wht WM
CAS 122-39-4

Specifications

Chemical Name or Material Diphenylamine
CAS 122-39-4
CAS Min % 98.0
CAS Max % 100.0
Purity Grade ACS Grade
Accessories

60 months