Please login to your online account to display your discounted pricing

Diphenylamine, ACS Grade, LabChem™

$98.63 - $98.63

Chemical Identifiers

Chemical Name or Material Diphenylamine
CAS 122-39-4
CAS Max % 100
Purity Grade ACS Grade
Linear Formula C12H11N
Formula Weight 169.22
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC136109

 
lab chem
LC136109
Amber Glass 100g Each for $98.63
Description & Specifications

Specifications

Formula Weight 169.22
Color White or almost white
Purity Grade ACS Grade
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
Harmful if swallowed.
May cause damage to organs (liver, kidneys, blood) through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Identification Passes Test
CAS 122-39-4
Health Hazard 3 GHS P Statement
Do not breathe dust.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.
Packaging Amber Glass
Storage Information Room Temperature
CAS Max % 100
EINECS Number 204-539-4
Linear Formula C12H11N
Density 1.16g/cm3
Physical Form Solid
Flash Point 153°C
Chemical Name or Material Diphenylamine
Solubility Soluble in water
Quantity 100g
Sensitivity Passes Test
Boiling Point 302°C
Safety and Handling

Warning

GHS P Statement
Do not breathe dust.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful if swallowed.
May cause damage to organs (liver, kidneys, blood) through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.