missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diphenylamine, 99%, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 150741000

 View more versions of this product

Catalog No. AC150741000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

Diphenylamine
52.0°C to 54.0°C
302.0°C
Authentic
Glass bottle
(C6H5)2NH
12,174
diphenylamine, n-phenylbenzenamine, anilinobenzene, benzenamine, n-phenyl, n,n-diphenylamine, phenylaniline, scaldip, n-phenylbenzeneamine, big dipper, no-scald
DMBHHRLKUKUOEG-UHFFFAOYSA-N
N-phenylaniline
11487
169.23
Pure
122-39-4
Orange to White
152°C
98.5% min. (GC)
C12H11N
100 g
15,3347
Solubility in water: 0.05g/L. Other solubilities: soluble in alcohol and propyl alcohol, freely soluble in benzene, ether, glacial acetic, acid and carbon disulfide
C1=CC=C(C=C1)NC2=CC=CC=C2
169.23
CHEBI:4640
99%
Powder, Crystals or Chunks

Chemical Identifiers

122-39-4
169.23
diphenylamine, n-phenylbenzenamine, anilinobenzene, benzenamine, n-phenyl, n,n-diphenylamine, phenylaniline, scaldip, n-phenylbenzeneamine, big dipper, no-scald
CHEBI:4640
C1=CC=C(C=C1)NC2=CC=CC=C2
C12H11N
DMBHHRLKUKUOEG-UHFFFAOYSA-N
11487
N-phenylaniline
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

EINECSNumber : 204-539-4

Recommended Storage : This reagent discolors on exposure to light

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only