Please login to your online account to display your discounted pricing

Diphenylamine, 99%, pure, ACROS Organics™

$17.88 - $38.23

Chemical Identifiers

Chemical Name or Material Diphenylamine
CAS 122-39-4
Purity Grade Pure
Assay ≥98.5% (GC)
Assay Percent Range 99%
Molecular Formula C12H11N
Linear Formula (C6H5)2NH
Formula Weight 169.23
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC150741000

  SDS
fisher scientific acros
AC150741000
Glass bottle 100g Each for $38.23

AC150740050

  SDS
thermo fisher scientific
150740050
Glass bottle 5g Each for $17.88

AC150740051

  SDS
thermo fisher scientific
AC150740051
Plastic Bucket 5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Diphenylamine
CAS 122-39-4
Health Hazard 1 Danger
Infrared Spectrum Authentic
Packaging Glass bottle
Purity Grade Pure
Physical Form Powder, Crystals and/or Chunks
Assay ≥98.5% (GC)
Assay Percent Range 99%
Molecular Formula C12H11N
Linear Formula (C6H5)2NH
Formula Weight 169.23
Storage Information This reagent discolors on exposure to light
Melting Point 52° to 55°C
Color White to yellow-beige to orange
Quantity 100g

Indicator

Safety and Handling

Danger