Learn More
Diphenylamine, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1826536
Description
Diphenylamine is employed as harvest scald inhibitor for apples indoor drench treatments due to its antioxidant property. Alkyl derivatives of diphenylamine are used as anti-ozonants in the rubber manufacturing. It is used as lubricants and acts as a stabilizer for plastics. In the analytical chemistry, it is used to test the nitrate ion in association with ammonium chloride in sulfuric acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiphenylamine is employed as harvest scald inhibitor for apples indoor drench treatments due to its antioxidant property. Alkyl derivatives of diphenylamine are used as anti-ozonants in the rubber manufacturing. It is used as lubricants and acts as a stabilizer for plastics. In the analytical chemistry, it is used to test the nitrate ion in association with ammonium chloride in sulfuric acid.
Solubility
Soluble in ethanol, acetone, benzene, carbon tetrachloride, pyridine and ethyl acetate. Slightly soluble in chloroform.
Notes
Incompatible with strong acids, strong oxidizing agents. Keep the container tightly closed in a cool and well-ventilated area.
Specifications
Diphenylamine | |
52°C to 55°C | |
302°C | |
Weak | |
(C6H5)2NH | |
500 g | |
508755 | |
14,3317 | |
Soluble in ethanol,acetone,benzene,carbon tetrachloride,pyridine and ethyl acetate. Slightly soluble in chloroform. | |
C1=CC=C(C=C1)NC2=CC=CC=C2 | |
169.227 | |
CHEBI:4640 | |
≥98% |
122-39-4 | |
1.16 | |
152°C (305°F) | |
C12H11N | |
MFCD00003014 | |
UN3077 | |
Air and light sensitive | |
diphenylamine, n-phenylbenzenamine, anilinobenzene, benzenamine, n-phenyl, n,n-diphenylamine, phenylaniline, scaldip, n-phenylbenzeneamine, big dipper, no-scald | |
DMBHHRLKUKUOEG-UHFFFAOYSA-N | |
N-phenylaniline | |
11487 | |
169.23 |
Chemical Identifiers
122-39-4 | |
169.227 | |
DMBHHRLKUKUOEG-UHFFFAOYSA-N | |
11487 | |
N-phenylaniline |
C12H11N | |
MFCD00003014 | |
diphenylamine, n-phenylbenzenamine, anilinobenzene, benzenamine, n-phenyl, n,n-diphenylamine, phenylaniline, scaldip, n-phenylbenzeneamine, big dipper, no-scald | |
CHEBI:4640 | |
C1=CC=C(C=C1)NC2=CC=CC=C2 |
Safety and Handling
GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c
H301+H311+H331-H373
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 204-539-4
RTECSNumber : JJ7800000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only