Please login to your online account to display your discounted pricing

Diphenylamine, 1% (w/v) Aqueous, Ricca Chemical

$123.42 - $123.42

Chemical Identifiers

Chemical Name or Material Diphenylamine
CAS 7732-18-5
CAS Min % 97.0
CAS Max % 100.0
CAS 6152-67-6
CAS Min % 0.97
CAS Max % 1.01
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

258716

 
ricca chemical company
RI258716
Poly natural 500mL Each for $123.42

25874

 
ricca chemical company
RI25874
Poly natural 120mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 500mL
Odor Odorless
Color Clear colorless
CAS Min % 97.0
CAS Max % 100.0
Chemical Name or Material Diphenylamine
Shelf Life 12 months
Packaging Poly natural
CAS 7732-18-5
Concentration or Composition (by Analyte or Components) 1% (w/v) Aqueous
CAS 6152-67-6
CAS Min % 0.97
CAS Max % 1.01
Safety and Handling

12 months