Please login to your online account to display your discounted pricing

Diphenylamine, 0.3% in 50% Sulfuric Acid, Certified, LabChem™

$151.43 - $151.43

Chemical Identifiers

Chemical Name or Material Diphenylamine
CAS 7664-93-9
CAS Max % 61.5
CAS 7732-18-5
CAS Max % 38.2
CAS 122-39-4
CAS Max % 0.3
Name Note 0.3% in 50% Sulfuric Acid
Purity Grade Certified Grade
Linear Formula C12H11N
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC136401

 
lab chem
LC136401
Amber Glass 500mL Each for $151.43
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 169.22
Chemical Name or Material Diphenylamine
CAS Max % 0.3
Identification Passes Test
Name Note 0.3% in 50% Sulfuric Acid
Packaging Amber Glass
Physical Form Liquid
Linear Formula C12H11N
Solubility Soluble in water
CAS 7664-93-9
Storage Information Room Temperature
UN Number UN1830
Health Hazard 1 Danger
Color Clear, colorless to slightly tan
Quantity 500mL
CAS Max % 61.5
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
CAS 7732-18-5
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Decomposition Information Sulfur compounds; Carbon monoxide; Carbon dioxide
Specific Gravity 1.51g/mL
CAS Max % 38.2
Density 1.51g/mL
Suitability Passes Test
CAS 122-39-4
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.