Please login to your online account to display your discounted pricing

Alfa Aesar™ Diphenylacetylene-4,4'-diboronic acid bis(pinacol) ester, 95%

$136.97 - $540.93

Chemical Identifiers

Chemical Name or Material Diphenylacetylene-4,4'-diboronic acid bis(pinacol) ester
CAS 849681-64-7
Molecular Formula C26H32B2O4
Formula Weight 430.16
MDL Number MFCD16294524
Synonym 4,4'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)diphenylacetylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5189703

  SDS
alfa aesar™
H5189703
1g Each for $136.97

AAH5189706

  SDS
alfa aesar™
H5189706
5g Each for $540.93
Description & Specifications

Specifications

Formula Weight 430.16
MDL Number MFCD16294524
Synonym 4,4'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)diphenylacetylene
Quantity 1g
Molecular Formula C26H32B2O4
Assay 95%
CAS 849681-64-7
Chemical Name or Material Diphenylacetylene-4,4'-diboronic acid bis(pinacol) ester

Applications
It is used as an active pharmaceutical intermediate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.