missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diphenylacetyl chloride, 97.5%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 292020250

 View more versions of this product

Catalog No. AC292020250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Diphenylacetyl chloride
Glass bottle
51°C to 56°C
97.5%
97.5%
(C6H5)2CHCOCl
09, 674
a-phenylbenzeneacetyl chloride, acetyl chloride, diphenyl, alpha-phenylbenzeneacetyl chloride, benzeneacetyl chloride, .alpha.-phenyl, diphenyl acetyl chloride, diphenyl-acetyl chloride, diphenylacetic acid chloride, diphenylacetyl chloride, diphenylacetylchloride, unii-6553c1cb9s
C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)Cl
230.691
230.69
≥96.5%
Authentic
Yellow
25g
1871-76-7
C14H11ClO
MFCD00013655
15, 3346
MSYLETHDEIJMAF-UHFFFAOYSA-N
2,2-diphenylacetyl chloride
74637
Crystalline Powder

Chemical Identifiers

1871-76-7
230.691
MSYLETHDEIJMAF-UHFFFAOYSA-N
74637
C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)Cl
C14H11ClO
MFCD00013655
a-phenylbenzeneacetyl chloride, acetyl chloride, diphenyl, alpha-phenylbenzeneacetyl chloride, benzeneacetyl chloride, .alpha.-phenyl, diphenyl acetyl chloride, diphenyl-acetyl chloride, diphenylacetic acid chloride, diphenylacetyl chloride, diphenylacetylchloride, unii-6553c1cb9s
2,2-diphenylacetyl chloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 217-493-5

RTECSNumber : AO6750000

SDS Documents