missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diphenylacetonitrile, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0240436

 View more versions of this product

Catalog No. AAL0240436



Description

Description

Diphenylacetonitrile undergoes anhydrous condensation with ethyl-4-bromo-butyrate. This is followed by its hydrolysis in alkaline medium that is used in the quantitative determination of 3-cyano-3,3-diphenylpropionic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylacetonitrile undergoes anhydrous condensation with ethyl-4-bromo-butyrate. This is followed by its hydrolysis in alkaline medium that is used in the quantitative determination of 3-cyano-3,3-diphenylpropionic acid.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

Diphenylacetonitrile
72°C to 76°C
120°C (248°F)
MFCD00001862
1911160
diphenylacetonitrile, dipan, diphenatrile, benzhydrylcyanide, diphenylmethylcyanide, benzyhydrylcyanide, acetonitrile, diphenyl, benzeneacetonitrile, alpha-phenyl, usaf kf-13, difenylacetonitril
NEBPTMCRLHKPOB-UHFFFAOYSA-N
2,2-diphenylacetonitrile
6837
99%
86-29-3
181°C (12 mmHg)
C14H11N
500 g
14,3316
Insoluble in water.
C1=CC=C(C=C1)C(C#N)C2=CC=CC=C2
193.249
193.25

Chemical Identifiers

86-29-3
193.249
NEBPTMCRLHKPOB-UHFFFAOYSA-N
6837
C1=CC=C(C=C1)C(C#N)C2=CC=CC=C2
C14H11N
MFCD00001862
diphenylacetonitrile, dipan, diphenatrile, benzhydrylcyanide, diphenylmethylcyanide, benzyhydrylcyanide, acetonitrile, diphenyl, benzeneacetonitrile, alpha-phenyl, usaf kf-13, difenylacetonitril
2,2-diphenylacetonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H315-H319-H335

EINECSNumber : 201-662-5

RTECSNumber : AL9800000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only