Learn More
Diphenylacetic Acid, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 117155000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diphenylacetic acid | |
117-34-0 | |
Cream-White to White | |
Authentic | |
Plastic bottle | |
(C6H5)2CHCO2H | |
500 g | |
15, 3346 | |
Solubility in water: soluble in hot water. Other solubilities: soluble in alcohol, ether and chloroform | |
C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)O | |
212.248 | |
CHEBI:41967 | |
≥99% |
99+% | |
146.0°C to 149.0°C | |
0.5% max. | |
99+% | |
C14H12O2 | |
MFCD00004251 | |
09, 673 | |
diphenylacetic acid, diphenylethanoic acid, acetic acid, diphenyl, benzeneacetic acid, .alpha.-phenyl, 1,1-diphenylacetic acid, unii-658ncz0nko, diphenyl-acetic acid, alpha-toluic acid, alpha-phenyl, 658ncz0nko, benzeneacetic acid, alpha-phenyl | |
PYHXGXCGESYPCW-UHFFFAOYSA-N | |
2,2-diphenylacetic acid | |
8333 | |
212.25 | |
Crystalline Powder |
Chemical Identifiers
117-34-0 | |
PYHXGXCGESYPCW-UHFFFAOYSA-N | |
8333 | |
2,2-diphenylacetic acid |
MFCD00004251 | |
diphenylacetic acid, diphenylethanoic acid, acetic acid, diphenyl, benzeneacetic acid, .alpha.-phenyl, 1,1-diphenylacetic acid, unii-658ncz0nko, diphenyl-acetic acid, alpha-toluic acid, alpha-phenyl, 658ncz0nko, benzeneacetic acid, alpha-phenyl | |
CHEBI:41967 | |
C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)O |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 204-185-0
RTECSNumber : AH2515000
TSCA : TSCA
RUO – Research Use Only