Please login to your online account to display your discounted pricing

Diphenylacetaldehyde, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC117140250

 View more versions of this product

Catalog No. AC117140250

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥96.0%
Formula Weight 196.25
Quantity 25g
Color Clear colorless to pale yellow
Beilstein 07, 438
Density 1.1g/mL
Chemical Name or Material Diphenylacetaldehyde
Molecular Formula C14H12O
Linear Formula (C6H5)2CHCHO
Assay Percent Range 97%
CAS 947-91-1
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5900 to 1.5920 (20°C, 589nm)
MDL Number MFCD00006972

Specifications

Chemical Name or Material Diphenylacetaldehyde
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C14H12O
Linear Formula (C6H5)2CHCHO
Formula Weight 196.25
MDL Number MFCD00006972
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents