Please login to your online account to display your discounted pricing

Diphenylacetaldehyde, 97%, ACROS Organics™

$339.22 - $339.22

Chemical Identifiers

Chemical Name or Material Diphenylacetaldehyde
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C14H12O
Linear Formula (C6H5)2CHCHO
Formula Weight 196.25
MDL Number MFCD00006972
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC117140250

  SDS
thermo fisher scientific
AC117140250
Glass bottle 25g Each for $339.22

AC117140050

  SDS
thermo fisher scientific
AC117140050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 196.25
Assay ≥96.0%
Quantity 25g
Color Clear colorless to pale yellow
Beilstein 07, 438
Density 1.1g/mL
Linear Formula (C6H5)2CHCHO
Molecular Formula C14H12O
Chemical Name or Material Diphenylacetaldehyde
CAS 947-91-1
Assay Percent Range 97%
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00006972
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.5900 to 1.5920 (20°C, 589nm)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.