Please login to your online account to display your discounted pricing

Alfa Aesar™ Diphenylacetaldehyde, 95%

Chemical Identifiers

Chemical Name or Material Diphenylacetaldehyde
CAS 947-91-1
Formula Weight 196.24g/mol
MDL Number MFCD00006972
Beilstein 1424292
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5654706

 
alfa aesar™
H56547-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5654714

 
alfa aesar™
H56547-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Density 1.106g/mL
Refractive Index 1.589
Boiling Point 315°C
Color Colorless to yellow
Quantity 5g
Chemical Name or Material Diphenylacetaldehyde
CAS 947-91-1
Physical Form Liquid
Assay 95%
Formula Weight 196.24g/mol
MDL Number MFCD00006972
Beilstein 1424292
RTECS Number AB2827500