Please login to your online account to display your discounted pricing

Alfa Aesar™ Diphenyl terephthalate, 97%

$100.51 - $100.51

Chemical Identifiers

Chemical Name or Material Diphenyl terephthalate
CAS 1539-04-4
Molecular Formula C20H14O4
Formula Weight 318.33
MDL Number MFCD00016574
Beilstein 1889204
Synonym Terephthalic acid diphenyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0287706

  SDS
alfa aesar™
L0287706
5g Each for $100.51
Description & Specifications

Specifications

Formula Weight 318.33
Chemical Name or Material Diphenyl terephthalate
TSCA TSCA
Assay 97%
Molecular Formula C20H14O4
CAS 1539-04-4
Synonym Terephthalic acid diphenyl ester
Odor Odorless
MDL Number MFCD00016574
Beilstein 1889204
Melting Point 196° to 198°C
Quantity 5g