Learn More
Diphenyl sulfoxide, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2213536
Description
Diphenyl sulfoxide is a reagent used as an oxidant and hydroxyl activator in combination with a variety of electrophilic reagents. Efficient glycosidation of a phenyl thiosialoside donor with diphenyl sulfoxide and triflic anhydride in dichloromethane was reported. Direct glycosylations with 1-hydroxy glycosyl donors is made using trifluoromethanesulfonic anhydride and diphenyl sulfoxide. The preparation of triarylsulfonium halides was by the action of aryl Grignard reagents on diphenyl sulfoxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiphenyl sulfoxide is a reagent used as an oxidant and hydroxyl activator in combination with a variety of electrophilic reagents. Efficient glycosidation of a phenyl thiosialoside donor with diphenyl sulfoxide and triflic anhydride in dichloromethane was reported. Direct glycosylations with 1-hydroxy glycosyl donors is made using trifluoromethanesulfonic anhydride and diphenyl sulfoxide. The preparation of triarylsulfonium halides was by the action of aryl Grignard reagents on diphenyl sulfoxide.
Solubility
Soluble in water. Soluble in most organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Diphenyl sulfoxide | |
69°C to 72°C | |
C12H10OS | |
500 g | |
diphenyl sulfoxide, phenyl sulfoxide, sulfinyldibenzene, phenylsulfinylbenzene, diphenyl sulphoxide, sulfoxide, diphenyl, diphenylsulfoxide, benzene, 1,1'-sulfinylbis, 1,1'-sulfinyldibenzene, phenylsulfinyl benzene | |
JJHHIJFTHRNPIK-UHFFFAOYSA-N | |
benzenesulfinylbenzene | |
13679 | |
≥98% |
945-51-7 | |
206°C to 208°C (13 mmHg) | |
MFCD00002085 | |
1908444 | |
Soluble in water. Soluble in most organic solvents. | |
C1=CC=C(C=C1)S(=O)C2=CC=CC=C2 | |
202.271 | |
202.28 |
Chemical Identifiers
945-51-7 | |
202.271 | |
JJHHIJFTHRNPIK-UHFFFAOYSA-N | |
13679 | |
C1=CC=C(C=C1)S(=O)C2=CC=CC=C2 |
C12H10OS | |
MFCD00002085 | |
diphenyl sulfoxide, phenyl sulfoxide, sulfinyldibenzene, phenylsulfinylbenzene, diphenyl sulphoxide, sulfoxide, diphenyl, diphenylsulfoxide, benzene, 1,1'-sulfinylbis, 1,1'-sulfinyldibenzene, phenylsulfinyl benzene | |
benzenesulfinylbenzene |
Safety and Handling
EINECSNumber : 213-415-9
RTECSNumber : DA9185000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only