missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Diphenyl sulfide, 98%

Manufacturer:  Alfa Aesar™ A1111422

 View more versions of this product

Catalog No. AAA1111422



Description

Description

Applications
Diphenyl sulfide is used as a raw material for triaryl sulfonium photoinitiator, intermediates for agrichemicals, catalyst, metal extractants.

Solubility
Soluble in ether, benzene, carbon disulphide.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents, acids, acid chlorides, acid anhydrides.
Specifications

Specifications

Diphenyl sulfide
98%
MFCD00003064
14,7314
LTYMSROWYAPPGB-UHFFFAOYSA-N
phenylsulfanylbenzene
8766
186.28
139-66-2
C12H10S
1907932
Phenyl sulfide
C1=CC=C(C=C1)SC2=CC=CC=C2
186.272
CHEBI:38959

Chemical Identifiers

139-66-2
186.272
LTYMSROWYAPPGB-UHFFFAOYSA-N
8766
phenylsulfanylbenzene
C12H10S
MFCD00003064
Phenyl sulfide
CHEBI:38959
C1=CC=C(C=C1)SC2=CC=CC=C2
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.

Warning

RTECSNumber : SX2275000

TSCA : TSCA

SDS Documents