Please login to your online account to display your discounted pricing

Alfa Aesar™ Diphenyl-n-propylphosphine, 97%

$27.12 - $259.16

Chemical Identifiers

Chemical Name or Material Diphenyl-n-propylphosphine
CAS 7650-84-2
Molecular Formula C15H17P
Formula Weight 228.28
MDL Number MFCD00015216
Beilstein 642795
Synonym n-Propyldiphenylphosphine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0240203

  SDS
alfa aesar™
L0240203
1g Each for $27.12

AAL0240206

  SDS
alfa aesar™
L0240206
5g Each for $76.39

AAL0240214

  SDS
alfa aesar™
L0240214
25g Each for $259.16
Description & Specifications

Specifications

Formula Weight 228.28
CAS 7650-84-2
Chemical Name or Material Diphenyl-n-propylphosphine
Health Hazard 2 GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
MDL Number MFCD00015216
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Flash Point >110°C (230°F)
Boiling Point 163° to 164°C (14mmHg)
Assay 97%
Molecular Formula C15H17P
Health Hazard 1 Warning
Synonym n-Propyldiphenylphosphine
Beilstein 642795
Quantity 1g
Odor Unpleasant
RTECS Number SY9150000
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.