Learn More
Diphenyl N-cyanocarbonimidate, 97%, Thermo Scientific Chemicals
$285.92 - $285.92
Chemical Identifiers
CAS | 79463-77-7 |
---|---|
Molecular Formula | C14H10N2O2 |
Molecular Weight (g/mol) | 238.25 |
MDL Number | MFCD00010380 |
InChI Key | SLIKWVTWIGHFJE-UHFFFAOYSA-N |
Synonym | diphenyl n-cyanocarbonimidate, diphenyl cyanocarbonimidate, diphenoxymethylenecyanamide, n-cyano-diphenyl imidocarbonate, diphenylcyanocarbonimidate, cyanocarbonimidia acid diphenyl aester, n-cyanocarbonimidic acid diphenyl ester, bis phenoxy methylidenecyanamide, cyano diphenoxymethylidene amine, 3,3-diphenoxy-2-azaprop-2-enenitrile |
PubChem CID | 688090 |
SMILES | N#CN=C(OC1=CC=CC=C1)OC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC297210100
|
Thermo Scientific Chemicals
297210100 |
10 g | Glass bottle |
Each for $285.92
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
79463-77-7 | |
238.25 | |
SLIKWVTWIGHFJE-UHFFFAOYSA-N | |
688090 |
C14H10N2O2 | |
MFCD00010380 | |
diphenyl n-cyanocarbonimidate, diphenyl cyanocarbonimidate, diphenoxymethylenecyanamide, n-cyano-diphenyl imidocarbonate, diphenylcyanocarbonimidate, cyanocarbonimidia acid diphenyl aester, n-cyanocarbonimidic acid diphenyl ester, bis phenoxy methylidenecyanamide, cyano diphenoxymethylidene amine, 3,3-diphenoxy-2-azaprop-2-enenitrile | |
N#CN=C(OC1=CC=CC=C1)OC1=CC=CC=C1 |
Specifications
79463-77-7 | |
White | |
96% min | |
C14H10N2O2 | |
MFCD00010380 | |
diphenyl n-cyanocarbonimidate, diphenyl cyanocarbonimidate, diphenoxymethylenecyanamide, n-cyano-diphenyl imidocarbonate, diphenylcyanocarbonimidate, cyanocarbonimidia acid diphenyl aester, n-cyanocarbonimidic acid diphenyl ester, bis phenoxy methylidenecyanamide, cyano diphenoxymethylidene amine, 3,3-diphenoxy-2-azaprop-2-enenitrile | |
SLIKWVTWIGHFJE-UHFFFAOYSA-N | |
cyano(diphenoxymethylidene)amine | |
688090 | |
97% | |
Diphenyl N-cyanocarbonimidate |
155.0°C to 160.0°C | |
Authentic | |
Glass bottle | |
NCN=C(OC6H5)2 | |
10 g | |
Solubility in water: may decompose. | |
N#CN=C(OC1=CC=CC=C1)OC1=CC=CC=C1 | |
238.25 | |
238.25 | |
Fluffy Powder |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO – Research Use Only