Please login to your online account to display your discounted pricing

Alfa Aesar™ Diphenyl methylphenyl phosphate, mixture of isomers, 94%

Chemical Identifiers

Chemical Name or Material Diphenyl methylphenyl phosphate
CAS 26444-49-5
Name Note Mixture of isomers
MDL Number MFCD00014920
UN Number UN3082
Synonym Phosphoric acid diphenyl methylphenyl ester; Cresyl diphenyl phosphate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42092AE

 
alfa aesar™
42092AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA42092AP

 
alfa aesar™
42092AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Diphenyl methylphenyl phosphate
TSCA TSCA
Refractive Index 1.5630
Name Note Mixture of isomers
Physical Form Liquid
Assay 94%
MDL Number MFCD00014920
CAS 26444-49-5
UN Number UN3082
Synonym Phosphoric acid diphenyl methylphenyl ester; Cresyl diphenyl phosphate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
RTECS Number TC5520000
Density 1.21g/mL
Flash Point 232° (450°F)
Boiling Point 390°C
Color Pale yellow
Melting Point −38°C
Quantity 100mL
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.