missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diphenyl Disulfide, TCI America™

Manufacturer:  TCI America P0167500G

 View more versions of this product

Catalog No. P0167500G



Specifications

Specifications

Diphenyl Disulfide
61°C
882-33-7
MFCD00003065
diphenyl disulfide, phenyl disulfide, disulfide, diphenyl, diphenyldisulfide, 1,2-diphenyldisulfane, phenyldisulfanyl benzene, diphenyl disulphide, biphenyl disulfide, disulfide diphenyl, phenyldithiobenzene
C1=CC=C(C=C1)SSC2=CC=CC=C2
218.332
218.33
≥99.0% (GC)
White-Yellow
500g
C12H10S2
3077
GUUVPOWQJOLRAS-UHFFFAOYSA-N
(phenyldisulfanyl)benzene
13436
Crystal-Powder at 20°C

Chemical Identifiers

882-33-7
218.332
GUUVPOWQJOLRAS-UHFFFAOYSA-N
13436
C1=CC=C(C=C1)SSC2=CC=CC=C2
C12H10S2
MFCD00003065
diphenyl disulfide, phenyl disulfide, disulfide, diphenyl, diphenyldisulfide, 1,2-diphenyldisulfane, phenyldisulfanyl benzene, diphenyl disulphide, biphenyl disulfide, disulfide diphenyl, phenyldithiobenzene
(phenyldisulfanyl)benzene
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1124

RTECSNumber : SS6825000

TSCA : Yes

Documents