Learn More
Diphenyl diselenide, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 177570250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diphenyl diselenide | |
60.0°C to 63.0°C | |
Authentic | |
Glass bottle | |
(C6H5)2Se2 | |
25 g | |
05,272; 06,235; 07,136; 09,199; 10,170; 11,219; 13,125; 16,152; 17,352 | |
Solubility in water: insoluble in water. | |
[Se]([Se]C1=CC=CC=C1)C1=CC=CC=C1 | |
312.15 | |
312.13 | |
Crystalline Powder |
1666-13-3 | |
Yellow | |
98.5% min. (GC) | |
C12H10Se2 | |
MFCD00003001 | |
06, 346 | |
diphenyl diselenide, diselenide, diphenyl, phenyl diselenide, 1,2-diphenyldiselane, diphenyldiselenide, bis phenylselenide, phenyldiselanyl benzene, diphenyldiselane, ccris 9227, acmc-209duq | |
YWWZCHLUQSHMCL-UHFFFAOYSA-N | |
(phenyldiselanyl)benzene | |
15460 | |
99% |
Chemical Identifiers
1666-13-3 | |
312.15 | |
YWWZCHLUQSHMCL-UHFFFAOYSA-N | |
15460 | |
[Se]([Se]C1=CC=CC=C1)C1=CC=CC=C1 |
C12H10Se2 | |
MFCD00003001 | |
diphenyl diselenide, diselenide, diphenyl, phenyl diselenide, 1,2-diphenyldiselane, diphenyldiselenide, bis phenylselenide, phenyldiselanyl benzene, diphenyldiselane, ccris 9227, acmc-209duq | |
(phenyldiselanyl)benzene |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and
GHS Signal Word: Danger
EINECSNumber : 216-780-2
RUO – Research Use Only