Please login to your online account to display your discounted pricing

Alfa Aesar™ Diphenyl cyanocarbonimidate, 97%

$51.23 - $632.84

Chemical Identifiers

Chemical Name or Material Diphenyl cyanocarbonimidate
CAS 79463-77-7
Molecular Formula C14H10N2O2
Linear Formula (C6H5O)2C=NCN
Formula Weight 238.25
MDL Number MFCD00010380
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1660003

 
alfa aesar™
A1660003
1g Each for $51.23

AAA1660006

 
alfa aesar™
A1660006
5g Each for $144.80

AAA1660014

 
alfa aesar™
A1660014
25g Each for $632.84
Description & Specifications

Specifications

Formula Weight 238.25
Chemical Name or Material Diphenyl cyanocarbonimidate
Assay 97%
Molecular Formula C14H10N2O2
Linear Formula (C6H5O)2C=NCN
MDL Number MFCD00010380
CAS 79463-77-7
Melting Point 155° to 158°C
Health Hazard 1 Danger
Quantity 1g
Health Hazard 2 GHS H Statement
H318
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280i-P273-P305+P351+P338
Wear eye/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Applications
Diphenyl N-cyanocarbonimidate has been used in the preparation of N-substituted 2-amino-4H-3,1-benzoxazines, cyanoguanidine derivatives, oxadiazoles.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep under a nitrogen blanket. Store away from strong oxidizing agents, strong acids, strong bases, moist air, water.

Safety and Handling

Danger

GHS P Statement
P280i-P273-P305+P351+P338
Wear eye/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H318
Causes serious eye damage.