missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diphenyl carbonate, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC117230100
Specifications
Diphenyl carbonate | |
Authentic | |
301.0°C to 302.0°C | |
78.0°C to 81.0°C | |
102-09-0 | |
C13H10O3 | |
06,158 | |
carbonic acid diphenyl ester, carbonic acid, diphenyl ester, diphenylcarbonate, ph2co3, phenyl carbonate, phenyl carbonate pho 2co, phenyl phenoxyformate, pho 2co, unii-ywv401idyn, ywv401idyn | |
ROORDVPLFPIABK-UHFFFAOYSA-N | |
diphenyl carbonate | |
7597 | |
214.22 | |
99% |
168°C | |
Plastic drum | |
White | |
10kg | |
98.5% min. (GC) | |
(C6H5O)2CO | |
15,3354 | |
Solubility in water: insoluble. Other solubilities: solulbe in acetone, hot alcohol, benzene, carbon, tetrachloride, ether, glacial acetic acid and, other organic solvents | |
C1=CC=C(C=C1)OC(=O)OC2=CC=CC=C2 | |
214.22 | |
CHEBI:34722 | |
Flakes or Crystalline Powder |
Chemical Identifiers
102-09-0 | |
214.22 | |
carbonic acid diphenyl ester, carbonic acid, diphenyl ester, diphenylcarbonate, ph2co3, phenyl carbonate, phenyl carbonate pho 2co, phenyl phenoxyformate, pho 2co, unii-ywv401idyn, ywv401idyn | |
CHEBI:34722 | |
C1=CC=C(C=C1)OC(=O)OC2=CC=CC=C2 |
C13H10O3 | |
ROORDVPLFPIABK-UHFFFAOYSA-N | |
7597 | |
diphenyl carbonate |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid release to the environment.
GHS Signal Word: Warning
EINECSNumber : 203-005-8