Learn More
Diphenyl carbonate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A123620B
Description
Used for synthesis of many important organic compounds. As agrochemical intermediates, Dyestuff intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed for synthesis of many important organic compounds. As agrochemical intermediates, Dyestuff intermediate.
Solubility
Soluble in ethanol, diethyl ether, carbon tetrachloride, acetic acid. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Diphenyl carbonate | |
76°C to 81°C | |
301°C to 302°C | |
Mild | |
MFCD00003037 | |
1074863 | |
carbonic acid, diphenyl ester, phenyl carbonate, diphenylcarbonate, carbonic acid diphenyl ester, phenyl carbonate pho 2co, unii-ywv401idyn, ph2co3, pho 2co, ywv401idyn, phenyl phenoxyformate | |
ROORDVPLFPIABK-UHFFFAOYSA-N | |
diphenyl carbonate | |
7597 | |
214.22 |
102-09-0 | |
1.3 | |
168°C (334°F) | |
C13H10O3 | |
1000 g | |
14,3602 | |
Soluble in ethanol,diethyl ether,carbon tetrachloride,acetic acid. Insoluble in water. | |
C1=CC=C(C=C1)OC(=O)OC2=CC=CC=C2 | |
214.22 | |
CHEBI:34722 | |
99% |
Chemical Identifiers
102-09-0 | |
214.22 | |
ROORDVPLFPIABK-UHFFFAOYSA-N | |
7597 | |
diphenyl carbonate |
C13H10O3 | |
MFCD00003037 | |
carbonic acid, diphenyl ester, phenyl carbonate, diphenylcarbonate, carbonic acid diphenyl ester, phenyl carbonate pho 2co, unii-ywv401idyn, ph2co3, pho 2co, ywv401idyn, phenyl phenoxyformate | |
CHEBI:34722 | |
C1=CC=C(C=C1)OC(=O)OC2=CC=CC=C2 |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 203-005-8
RTECSNumber : FG0500000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only